Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142265.2 | XP_014997751.1 | 1516 | 90 | ||
XM_015142266.2 | XP_014997752.1 | 1475 | 90 | |||
XM_015142267.2 | XP_014997753.1 | 1632 | 90 | |||
XM_015142268.2 | XP_014997754.1 | 1475 | 90 | |||
XM_015142269.2 | XP_014997755.1 | 1475 | 90 | |||
XM_015142270.2 | XP_014997756.1 | 1632 | 90 | |||
XM_015142271.2 | XP_014997757.1 | 1475 | 90 | |||
XM_028850935.1 | XP_028706768.1 | 1475 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |