Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257433.1 | NP_001244362.1 | 580 | 89 | ||
XM_015125574.2 | XP_014981060.1 | 1831 | 89 | |||
XM_015125576.2 | XP_014981062.1 | 707 | 89 | |||
XM_015125578.2 | XP_014981064.1 | 645 | 89 | |||
XM_015125579.2 | XP_014981065.1 | 761 | 89 | |||
XM_028840530.1 | XP_028696363.1 | 586 | 89 | |||
XM_028840531.1 | XP_028696364.1 | 614 | 89 | |||
GenBank mRNA | JU321035.1 | - | 580 | 89 | ||
JU476037.1 | - | 564 | 89 | |||
JV045175.1 | - | 593 | 89 | |||
JV653914.1 | - | 595 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |