Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265864.1 | NP_001252793.1 | 796 | 100 | ||
GenBank mRNA | JU321606.1 | - | 842 | 100 | ||
JU473369.1 | - | 796 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |