Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032869.1 | NP_001028041.1 | 1003 | 102 | ||
XM_015111222.2 | XP_014966708.2 | 1037 | 102 | |||
XM_015111223.2 | XP_014966709.2 | 1156 | 102 | |||
GenBank mRNA | AF360121.1 | - | 1003 | 102 | ||
AY029169.1 | - | 1003 | 102 | |||
JU329361.1 | - | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta Inventoried |