Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841837.1 | XP_028697670.1 | 240 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |