Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032966.1 | NP_001028138.1 | 426 | 103 | ||
XM_015120877.2 | XP_014976363.2 | 1557 | 103 | |||
GenBank mRNA | JU322079.1 | - | 749 | 103 | ||
S78209.1 | - | 426 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Macaca mulatta |