Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266750.1 | NP_001253679.1 | 519 | 145 | ||
XM_015114235.2 | XP_014969721.2 | 684 | 145 | |||
XM_028827766.1 | XP_028683599.1 | 534 | 145 | |||
XM_028827863.1 | XP_028683696.1 | 595 | 145 | |||
GenBank mRNA | JU475325.1 | - | 505 | 145 | ||
JV046875.1 | - | 505 | 145 | |||
JV579744.1 | - | 689 | 145 | |||
JV636116.1 | - | 519 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |