Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001278456.1 | NP_001265385.1 | 3120 | 55 | ||
NM_001278457.1 | NP_001265386.1 | 3120 | 55 | |||
XM_028832502.1 | XP_028688335.1 | 2330 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |