Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2W | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266510.1 | NP_001253439.1 | 493 | 118 | ||
XM_015145506.2 | XP_015000992.1 | 546 | 118 | |||
XM_015145507.2 | XP_015000993.1 | 661 | 118 | |||
GenBank mRNA | JU320318.1 | - | 478 | 118 | ||
JU470972.1 | - | 470 | 118 | |||
JV047546.1 | - | 493 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |