Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104510.4 | XP_001104510.1 | 309 | 144 | ||
XM_015118004.2 | XP_014973490.2 | 372 | 144 | |||
XM_028835285.1 | XP_028691118.1 | 326 | 144 | |||
XM_028835289.1 | XP_028691122.1 | 235 | 144 | |||
XM_028835290.1 | XP_028691123.1 | 213 | 144 | |||
XM_028835291.1 | XP_028691124.1 | 198 | 144 | |||
XM_028835292.1 | XP_028691125.1 | 153 | 144 | |||
XM_028835293.1 | XP_028691126.1 | 168 | 144 | |||
GenBank mRNA | JU343656.1 | - | 155 | 144 | ||
JV582035.1 | - | 206 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |