Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266382.1 | NP_001253311.1 | 1143 | 94 | ||
XM_015110018.2 | XP_014965504.2 | 1414 | 94 | |||
XM_015110019.2 | XP_014965505.2 | 1344 | 94 | |||
XM_015110020.2 | XP_014965506.1 | 1243 | 94 | |||
GenBank mRNA | JU334320.1 | - | 1251 | 94 | ||
JU475422.1 | - | 1143 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |