Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266382.1 | NP_001253311.1 | 583 | 120 | ||
XM_015110020.2 | XP_014965506.1 | 683 | 120 | |||
GenBank mRNA | JU334320.1 | - | 691 | 120 | ||
JU475422.1 | - | 583 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |