Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPOPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266382.1 | NP_001253311.1 | 1088 | 75 | ||
XM_015110018.2 | XP_014965504.2 | 1359 | 75 | |||
XM_015110019.2 | XP_014965505.2 | 1289 | 75 | |||
XM_015110020.2 | XP_014965506.1 | 1188 | 75 | |||
GenBank mRNA | JU334320.1 | - | 1196 | 75 | ||
JU475422.1 | - | 1088 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |