Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCLAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104502.4 | XP_001104502.1 | 155 | 109 | ||
XM_015112040.2 | XP_014967526.1 | 150 | 109 | |||
GenBank mRNA | JV647302.1 | - | 91 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |