Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194345.1 | NP_001181274.1 | 535 | 88 | ||
XM_015150002.2 | XP_015005488.1 | 542 | 88 | |||
XM_028827380.1 | XP_028683213.1 | 488 | 88 | |||
GenBank mRNA | JU339678.1 | - | 977 | 88 | ||
JU473723.1 | - | 428 | 88 | |||
JV045029.1 | - | 429 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |