Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112823.2 | XP_014968309.2 | 2325 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |