Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPNMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266650.1 | NP_001253579.1 | 1598 | 131 | ||
XM_015133861.2 | XP_014989347.1 | 1751 | 131 | |||
XM_015133862.2 | XP_014989348.1 | 1840 | 131 | |||
XM_015133863.2 | XP_014989349.1 | 1752 | 131 | |||
XM_015133864.2 | XP_014989350.1 | 1541 | 131 | |||
GenBank mRNA | JU332515.1 | - | 1618 | 131 | ||
JU473990.1 | - | 1598 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |