Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLCXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260577.1 | NP_001247506.1 | 207 | 124 | ||
GenBank mRNA | JU322096.1 | - | 207 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |