Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMCES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194040.1 | NP_001180969.1 | 1068 | 102 | ||
XM_001095958.4 | XP_001095958.1 | 1025 | 102 | |||
XM_015130134.2 | XP_014985620.1 | 986 | 102 | |||
XM_028843211.1 | XP_028699044.1 | 1156 | 102 | |||
XM_028843212.1 | XP_028699045.1 | 978 | 102 | |||
GenBank mRNA | JU335538.1 | - | 895 | 102 | ||
JU476466.1 | - | 898 | 102 | |||
JV513579.1 | - | 852 | 102 | |||
JV587837.1 | - | 894 | 102 | |||
JV656419.1 | - | 799 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |