Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIWIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195743.1 | NP_001182672.1 | 2236 | 140 | ||
GenBank mRNA | HQ025804.1 | - | 2236 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |