Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194886.2 | NP_001181815.1 | 729 | 129 | ||
XM_001106204.3 | XP_001106204.1 | 910 | 129 | |||
XM_015141678.2 | XP_014997164.1 | 774 | 129 | |||
XM_028850397.1 | XP_028706230.1 | 1161 | 129 | |||
GenBank mRNA | JU321211.1 | - | 870 | 129 | ||
JU471015.1 | - | 870 | 129 | |||
JU471016.1 | - | 734 | 129 | |||
JV045757.1 | - | 866 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |