Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193769.1 | NP_001180698.1 | 204 | 240 | ||
XM_015143701.2 | XP_014999187.1 | 220 | 288 | |||
GenBank mRNA | JU320593.1 | - | 184 | 240 | ||
JU472064.1 | - | 155 | 240 | |||
JV045859.1 | - | 189 | 240 | |||
JV528696.1 | - | 168 | 240 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |