Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZC3H12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127584.2 | XP_014983070.1 | 2157 | 114 | ||
XM_015127585.2 | XP_014983071.1 | 2089 | 114 | |||
XM_015127587.2 | XP_014983073.1 | 2093 | 114 | |||
XM_028841848.1 | XP_028697681.1 | 2679 | 114 | |||
GenBank mRNA | JU324480.1 | - | 1052 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |