Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194394.2 | NP_001181323.1 | 1489 | 62 | ||
XM_015126510.2 | XP_014981996.1 | 1757 | 62 | |||
XM_028840541.1 | XP_028696374.1 | 1710 | 62 | |||
XM_028840543.1 | XP_028696376.1 | 2033 | 62 | |||
XM_028840544.1 | XP_028696377.1 | 1574 | 62 | |||
GenBank mRNA | JV044015.1 | - | 1525 | 62 | ||
JV635750.1 | - | 1495 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |