Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001089281.4 | XP_001089281.2 | 514 | 144 | ||
XM_028843647.1 | XP_028699480.1 | 523 | 144 | |||
GenBank mRNA | JU481097.1 | - | 473 | 144 | ||
JV643392.1 | - | 248 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |