Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135428.2 | XP_014990914.2 | 1112 | 73 | ||
XM_028847888.1 | XP_028703721.1 | 706 | 73 | |||
XM_028847889.1 | XP_028703722.1 | 1120 | 73 | |||
GenBank mRNA | JV059790.1 | - | 86 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |