Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYLK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135428.2 | XP_014990914.2 | 1565 | 124 | ||
XM_028847888.1 | XP_028703721.1 | 1159 | 124 | |||
XM_028847889.1 | XP_028703722.1 | 1573 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |