Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265620.1 | NP_001252549.1 | 97 | 76 | ||
GenBank mRNA | JV565002.1 | - | 97 | 76 | ||
JV602349.1 | - | 300 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |