Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099551.4 | XP_001099551.1 | 1525 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |