Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACYP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144143.1 | XP_014999629.1 | 335 | 124 | ||
XM_015144144.2 | XP_014999630.1 | 192 | 124 | |||
XM_015144145.2 | XP_014999631.1 | 225 | 124 | |||
XM_015144146.2 | XP_014999632.1 | 170 | 124 | |||
GenBank mRNA | JU473101.1 | - | 72 | 124 |
Gene Symbol: LOC697471 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001090451.4 | XP_001090451.1 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |