Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085299.4 | XP_001085299.2 | 425 | 103 | ||
XM_028841229.1 | XP_028697062.1 | 643 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |