Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTKN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261179.1 | NP_001248108.1 | 639 | 81 | ||
XM_015112538.2 | XP_014968024.1 | 2133 | 81 | |||
XM_015112539.2 | XP_014968025.1 | 1573 | 81 | |||
XM_015112540.2 | XP_014968026.1 | 1166 | 81 | |||
XM_015112541.2 | XP_014968027.1 | 1711 | 81 | |||
XM_015112543.2 | XP_014968029.1 | 1169 | 81 | |||
XM_028831552.1 | XP_028687385.1 | 1163 | 81 | |||
XM_028831553.1 | XP_028687386.1 | 3945 | 81 | |||
XM_028831554.1 | XP_028687387.1 | 2139 | 81 | |||
XM_028831555.1 | XP_028687388.1 | 1048 | 81 | |||
XM_028831559.1 | XP_028687392.1 | 2144 | 81 | |||
XM_028831560.1 | XP_028687393.1 | 1167 | 81 | |||
GenBank mRNA | JU321015.1 | - | 1068 | 81 | ||
JU322391.1 | - | 639 | 81 | |||
JV043526.1 | - | 1010 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |