Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137465.2 | XP_014992951.2 | 920 | 100 | ||
XM_015137466.2 | XP_014992952.2 | 729 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |