Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257623.3 | NP_001244552.1 | 1714 | 88 | ||
XM_015115699.2 | XP_014971185.2 | 1964 | 88 | |||
XM_015115700.2 | XP_014971186.2 | 1647 | 88 | |||
XM_028832514.1 | XP_028688347.1 | 1807 | 88 | |||
GenBank mRNA | JU320637.1 | - | 1717 | 88 | ||
JV528575.1 | - | 1718 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |