Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142361.2 | XP_014997847.1 | 2800 | 104 | ||
XM_015142362.2 | XP_014997848.1 | 2844 | 104 | |||
XM_015142364.2 | XP_014997850.1 | 2680 | 104 | |||
XM_015142365.2 | XP_014997851.1 | 2656 | 104 | |||
XM_015142366.2 | XP_014997852.1 | 2684 | 104 | |||
GenBank mRNA | JU322771.1 | - | 2606 | 104 | ||
JU477218.1 | - | 2784 | 104 | |||
JV723423.1 | - | 2596 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |