Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYZAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028850979.1 | XP_028706812.1 | 206 | 135 | ||
XM_028850980.1 | XP_028706813.1 | 206 | 135 | |||
GenBank mRNA | JU321531.1 | - | 189 | 135 | ||
JV704273.1 | - | 196 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |