Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FANCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126993.2 | XP_014982479.2 | 3542 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |