Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260979.1 | NP_001247908.1 | 848 | 146 | ||
XM_028840444.1 | XP_028696277.1 | 876 | 146 | |||
XM_028840445.1 | XP_028696278.1 | 876 | 146 | |||
XM_028840446.1 | XP_028696279.1 | 876 | 146 | |||
XM_028840447.1 | XP_028696280.1 | 876 | 146 | |||
GenBank mRNA | JU321125.1 | - | 845 | 146 | ||
JU473282.1 | - | 719 | 146 | |||
JV047305.1 | - | 848 | 146 | |||
JV653365.1 | - | 851 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |