Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266110.1 | NP_001253039.1 | 426 | 81 | ||
XM_015132019.2 | XP_014987505.1 | 282 | 81 | |||
XM_015132021.2 | XP_014987507.1 | 364 | 81 | |||
XM_015132023.2 | XP_014987509.1 | 224 | 81 | |||
XM_015132024.2 | XP_014987510.1 | 313 | 81 | |||
XM_028843395.1 | XP_028699228.1 | 285 | 81 | |||
XM_028843396.1 | XP_028699229.1 | 514 | 81 | |||
GenBank mRNA | JU322961.1 | - | 426 | 81 | ||
JU323054.1 | - | 366 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |