Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193798.1 | NP_001180727.1 | 682 | 113 | ||
GenBank mRNA | JU322588.1 | - | 657 | 113 | ||
JV043242.1 | - | 665 | 113 | |||
JV567600.1 | - | 686 | 113 | |||
JV602574.1 | - | 684 | 113 | |||
JV638267.1 | - | 676 | 113 | |||
JV669035.1 | - | 681 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |