Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTPAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265955.1 | NP_001252884.1 | 744 | 134 | ||
XM_015148844.2 | XP_015004330.1 | 1972 | 134 | |||
XM_015148845.2 | XP_015004331.1 | 1226 | 134 | |||
GenBank mRNA | JU336364.1 | - | 744 | 134 | ||
JU472731.1 | - | 743 | 134 | |||
JV723251.1 | - | 745 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |