Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257721.1 | NP_001244650.1 | 438 | 91 | ||
XM_015137591.2 | XP_014993077.1 | 1532 | 91 | |||
XM_028837599.1 | XP_028693432.1 | 1037 | 91 | |||
XM_028837665.1 | XP_028693498.1 | 1776 | 91 | |||
GenBank mRNA | JU335016.1 | - | 438 | 91 | ||
JU476021.1 | - | 500 | 91 | |||
JV047393.1 | - | 498 | 91 | |||
JV638729.1 | - | 515 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |