Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123706.2 | XP_014979192.2 | 411 | 67 | ||
XM_028839237.1 | XP_028695070.1 | 386 | 67 | |||
GenBank mRNA | JU480926.1 | - | 362 | 67 | ||
JV052676.1 | - | 363 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |