Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113649.4 | XP_001113649.2 | 881 | 80 | ||
GenBank mRNA | JV715281.1 | - | 251 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |