Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032807.2 | NP_001027979.1 | 300 | 143 | ||
XM_028846608.1 | XP_028702441.1 | 469 | 143 | |||
XM_028846610.1 | XP_028702443.1 | 469 | 143 | |||
GenBank mRNA | AF016372.1 | - | 303 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |