Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCN3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194283.2 | NP_001181212.1 | 978 | 152 | ||
XM_015115925.2 | XP_014971411.1 | 978 | 152 | |||
XM_028832733.1 | XP_028688566.1 | 740 | 152 | |||
GenBank mRNA | JU321695.1 | - | 1392 | 152 | ||
JU321697.1 | - | 993 | 152 | |||
JV601467.1 | - | 995 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |