Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261335.1 | NP_001248264.1 | 401 | 107 | ||
XM_015135553.2 | XP_014991039.1 | 739 | 107 | |||
GenBank mRNA | JU337113.1 | - | 456 | 107 | ||
JU473281.1 | - | 401 | 107 | |||
JV487261.1 | - | 347 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |