Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001083049.4 | XP_001083049.2 | 814 | 150 | ||
XM_015143179.2 | XP_014998665.2 | 317 | 150 | |||
GenBank mRNA | JU323393.1 | - | 797 | 150 | ||
JV473218.1 | - | 152 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |