Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100309.4 | XP_001100309.1 | 2105 | 119 | ||
XM_015135278.2 | XP_014990764.1 | 2077 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |