Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHI3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265920.1 | NP_001252849.1 | 181 | 135 | ||
GenBank mRNA | JU323002.1 | - | 181 | 135 | ||
JV043861.1 | - | 195 | 135 | |||
JV704305.1 | - | 202 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |